Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Fos Sheep anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1584MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1584MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1584MI Supplier MilliporeSigma Supplier No. AB1584
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Use Anti-c-Fos Antibody (Sheep Polyclonal Antibody) validated in immunohistochemistry, immunohistochemistry (paraffin), western blotting to detect c-Fos also known as G0/G1 Switch Regulatory Protein 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Fos
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Lyophilized, no preservatives. Reconstitute with 500μL of sterile distilled water (1mg/mL).
Gene Symbols FOS; G0S7; c-fos
Host Species Sheep
Immunogen Synthetic c-fos peptide (residues Ser.Gly.Phe.Asn.Ala.Asp.Tyr.Glu.Ala. Ser.Ser.Ser.Arg.Cys) corresponding to amino acids 4-17 of human Fos, conjugated to bovine thyroglobulin.
Purification Method Ammonium sulfate precipitation
Quantity 500 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005252.2
Target Species Rat
Content And Storage Lyophilized: −20-−70°C for up to 6 months. Reconstituted: −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.