Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
c-Fos Sheep anti-Human, Rat, Polyclonal, MilliporeSigma™
Sheep Polyclonal Antibody
Supplier: MilliporeSigma CBL440
Description
Anti-c-Fos Antibody is a Sheep Polyclonal Antibody for detection of c-Fos also known as G0/G1 Switch Regulatory Protein 7 and has been validated in western blot, immunocytochemistry.
Specifications
c-Fos | |
Polyclonal | |
The purified immunoglobulin is presented at a concentration of 100μg/ml in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application. | |
Sheep | |
100 μg | |
NM_005252.2 | |
4°C for 1 month, −20°C in undiluted aliquotes for long term |
Immunocytochemistry, Western Blot | |
Unconjugated | |
FOS; G0S7; c-fos | |
Synthetic peptide: Met-Phe-Ser-Gly-Phe-Asn-Ala-Asp-Tyr-Glu-Ala-Ser-Ser-Ser-Arg-Cys, selected from a conserved region of mouse and human c-fos. | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction