Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Fos Mouse anti-Human, Clone: 2G9C3, MilliporeSigma™

Catalog No. MABE329MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE329MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE329MI Supplier MilliporeSigma Supplier No. MABE329
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-c-Fos Antibody, clone 2G9C3 is a Mouse Monoclonal Antibody for detection of c-Fos also known as Proto-oncogene c-Fos, Cellular oncogene fos and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Fos
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2G9C3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ from cultured supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005243
Gene Symbols FOS, G0S7, c-Fos
Host Species Mouse
Immunogen Linear peptide corresponding to human c-Fos.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.