Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Jun Rabbit anti-Avian, Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06225MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-225-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-225-MI Supplier MilliporeSigma Supplier No. 06225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect c-Jun, also known as transcription factor AP-1, with Anti-c-Jun rabbit polyclonal antiserum, Cat. No. 06-225, that has been demonstrated to work in Immunocytochemistry, Immunoprecipitation, and Western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Jun
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation whole rabbit antiserum containing 0.05% sodium azide
Gene Symbols JUN; p39; AP1; c-Jun
Host Species Rabbit
Immunogen Trp E fusion-protein containing the DNA binding domain of avian c-Jun
Purification Method Antiserum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_010591
Target Species Avian, Human, Mouse
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.