Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Jun (pS63) Mouse, Unlabeled, Clone: 2/C-JUN (PS63), BD

Catalog No. BDB558036 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558036 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558036 Supplier BD Biosciences Supplier No. 558036
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The activator protein transcription factor (AP-1) was identified as a protein that recognizes specific sequences in the cis-control regions of the SV40 virus and the human metallothionein IIA gene. AP-1 is composed of protein products of two different gene families: jun and fos. The AP-1 transcription factor is either a homodimer of Jun proteins or a heterodimer of Jun and Fos proteins. The transcriptional activity of Jun is enhanced by phosphorylation of serines 63 (S63) and 73 in its activation domain. Phosphorylation at both sites is necessary for stimulating the activating function of Jun. Jun is phosphorylated by JNK protein kinases that are activated by the same signals that potentiate Jun activity. The 2/c-Jun/(S63) monoclonal antibody recognizes the phosphorylated S63 of c-Jun.

Host Species: Mouse
Clone: 2
Isotype: IgG1
Species Reactivity: Human

Immunohistochemistry (Formalin-Antigen Retrieval Required), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Jun (pS63)
Applications Western Blot
Classification Monoclonal
Clone 2/C-JUN (PS63)
Concentration 0.25mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human c-Jun
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.