Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Kit Mouse anti-Human, Clone: 1DC3, MilliporeSigma™

Catalog No. MAB1164MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1164MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1164MI Supplier MilliporeSigma Supplier No. MAB1164

Mouse Monoclonal Antibody

This Anti-c-Kit Antibody, clone 1DC3 is validated for use in flow cytometry, immunohistochemistry, immunoprecipitation, western blotting for the detection of c-Kit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Kit
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1DC3
Conjugate Unconjugated
Formulation Purified Immunoglobulin. Liquid in 0.02M phosphate buffer with 0.25MNaCl, pH=7.6 with 0.1% sodium Azide added as a preservative.
Gene Accession No. NM_000222.2; NM_001093772.1
Gene Symbols KIT, C-Kit, PBT, CD117, SCFR, c-kit, EC 2.7.10.1
Host Species Mouse
Immunogen Human erythroleukemia cell line.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.