Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ c-myc Mouse anti-Human, Mouse, Clone: 3C7

Catalog No. CBL434MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL434MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL434MI Supplier MilliporeSigma™ Supplier No. CBL434
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect c-myc also known as Proto-oncogene c-Myc with Anti-c-myc Antibody, clone 3C7 (Mouse Monoclonal Antibody), that has been demonstrated to work in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Myc
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3C7
Conjugate Unconjugated
Formulation The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular appli
Gene Accession No. NM_002467.3
Gene Symbols MYC, C-MYC, c-Myc
Host Species Mouse
Immunogen Synthetic peptide: Cys-Ser-Thr-Ser-Ser-Leu-Tyr-Leu-Gln-Asp-Leu-Ser-Ala-Ala-Ala-Ser-Glu-Cys selected from the human c-myc sequence (amino acids (171-188). The peptide was conjugated to keyhole limpet haemocyanin.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.