Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-c-Myc Clone: 90000000000,

Catalog No. OP107100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP107100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP107100UG Supplier MilliporeSigma™ Supplier No. OP107100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects c-Myc Clone: 90000000000 in Human, Mouse, Rat samples, and it is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting

The v-myc oncogene, initially identified in the MC29 avian retrovirus, causes myelocytomas, carcinomas, sarcomas and lymphomas, and belongs to a family of oncogenes conserved throughout evolution. In humans the family consists of five genes: c-myc, N-myc, R-myc, L-myc, and B-myc. Amplification of the c-myc gene has been found in several types of human tumors including lung, breast and colon carcinomas, while the N-myc gene has been found amplified in neuroblastomas, small cell lung carcinomas and Wilms tumors. L-myc is amplified in small cell lung cancer. Immunological studies have shown that the human c-myc gene gives rise to at least two nuclear phosphoproteins of ∽64kDa and ∽67kDa that exhibit relatively short (30 min) half lives in vivo and exhibit DNA binding properties in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Myc
Applications Flow Cytometry, Immunoblot, Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 90000000000
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen a synthetic peptide (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) corresponding to amino acids 408-439 of human c-Myc
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽60-67kDa c-Myc protein in HL60 cells and lung carcinoma tissue.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.