Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc with control Mouse anti-Human, Unlabeled, Clone: 9E10., BD

Catalog No. BDB551102 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551102 150 μg
BDB551101 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB551102 Supplier BD Biosciences Supplier No. 551102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The myc gene family contains at least seven closely related genes, c-myc, N-myc, L-myc, P-myc, R-myc, S-myc, and B-myc. C-myc plays a role in proliferation, transformation, and differentiation. It is expressed during embryonic development, in a wide variety of adult tissues, and is amplified in many tumors, particularly lung, breast, cervical and colon carcinomas. Cellular localization has been described as nuclear and/or cytopasmic. C-myc has three sequence motifs in the carboxy terminus, a leucine zipper, a helix-loop-helix, and a basic region. It forms sequence-specific (CACGTG) DNA binding heterodimers with max, a helixloop-helix/leucine zipper protein. Both the leucine zipper domain and the helixloop-helix motif of c-myc contribute to heterodimer formation. DNA binding of myc-max dimers results in a conformational change of the DNA and transcriptional activation. C-myc migrates at 62 kDa in SDS-PAGE. Clone 9E10 recognizes human c-myc. A synthetic peptide corresponding to a C-terminal epitope of the human c-myc protein (AEEQKLISEEDL) was used as an immunogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Myc with control
Applications Western Blot
Classification Monoclonal
Clone 9E10.
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Includes Purified Mouse Anti-Human c-Myc (150μg) 3vials of 50μg; Jurkat Cell Lysate (50μg)
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.