Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-myc Chicken anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3252MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3252MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3252MI Supplier MilliporeSigma Supplier No. AB3252
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-c-myc Antibody is a Chicken Polyclonal Antibody for detection of c-myc also known as Proto-oncogene c-Myc and has been tested in immunoprecipitation, western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-myc
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified chicken IgY. Liquid in PBS with 0.02% sodium azide.
Gene Symbols MYC; C-MYC; c-Myc
Host Species Chicken
Immunogen Synthetic peptide EQKLISEEDL from human c-myc.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002467.3
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.