Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-X-C Chemokine Receptor 4 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB1847MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1847MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1847MI Supplier MilliporeSigma Supplier No. AB1847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-C-X-C Chemokine Receptor 4 Antibody, extracellular loop detects level of C-X-C Chemokine Receptor 4 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-X-C Chemokine Receptor 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified IgG in PBS containing 0.02% sodium azide.
Gene Symbols CXCR4; WHIM; NPY3R; NPYY3R; FB22; LAP3; Fusin; CXCR-4; LCR1; NPYR; HM89; fusin; HSY3RR; CXC-R4; NPYY3; NPYRL; D2S201E; CD184; LESTR
Host Species Rabbit
Immunogen Extracellular loop peptide of human CXCR4 (Berson et al. 1996; Leotscher et al. 1994).
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001008540.1; NM_003467.2
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months in undiluted aliquotes.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.