Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-X-C Chemokine Receptor 4 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1846MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1846MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1846MI Supplier MilliporeSigma Supplier No. AB1846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-C-X-C Chemokine Receptor 4 Antibody, N-terminus is an antibody against C-X-C Chemokine Receptor 4 for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-X-C Chemokine Receptor 4
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified IgG in PBS containing 0.02% sodium azide.
Gene Symbols CXCR4; WHIM; NPY3R; NPYY3R; FB22; LAP3; Fusin; CXCR-4; LCR1; NPYR; HM89; fusin; HSY3RR; CXC-R4; NPYY3; NPYRL; D2S201E; CD184; LESTR
Host Species Rabbit
Immunogen Amino-terminus peptide of human CXCR4 (Berson, 1996; Leotscher, 1994).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001008540.1; NM_003467.2
Target Species Human
Content And Storage 4°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.