Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chromosome 10 open reading frame 46, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004795 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-795 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-795 Supplier Abnova Corporation Supplier No. H00143384A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant C10orf46.

Sequence: STININTSTSKFLMNVITIEDYKSTYWPKLDGAIDQLLTQSPGDYIPISYEQIYSCVYKCVCQQHSEQMYSDLIKKITNHLERVSKELQASPPDLYIERF

Specifications

Antigen chromosome 10 open reading frame 46
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant C10orf46.
Formulation 50% glycerol
Gene C10orf46
Gene Accession No. NM_153810
Gene Alias FLJ40409/MGC33215
Gene Symbols C10orf46
Host Species Mouse
Immunogen C10orf46 (NP_722517, 111 a.a. ∼ 210 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 143384
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.