Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C11orf30 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526485
Description
This antibody is predicted to react with mouse based on sequence homology.
Regulator which is able to repress transcription, possibly via its interaction with a multiprotein chromatin remodeling complex that modifies the chromatin. Its interaction with BRCA2 suggests that it may play a central role in the DNA repair function of BRCA2.
Specifications
C11orf30 | |
Polyclonal | |
EMSY | |
EMSY, protein EMSY | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56946 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q7Z589 | |
EMSY | |
KLH conjugated synthetic peptide between 46-75 amino acids from the N-terminal region of human C11orf30 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction