Learn More
chromosome 17 open reading frame 75, Mouse, Clone: 2D7, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | chromosome 17 open reading frame 75 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2D7 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant C17orf75. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | C17orf75 |
| Gene Accession No. | NM_022344 |
| Gene Alias | NJMU-R1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.