Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-C19orf50, Polyclonal, Abnova™

Catalog No. 89300496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-496 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-496 Supplier Abnova Corporation Supplier No. H00079036B01P
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

Specifications

Antigen C19orf50
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human C19orf50 protein.
Formulation PBS with no preservative; pH 7.4
Gene KXD1
Gene Accession No. NM_024069.2
Gene Alias FLJ25480/MGC2749/MST096/MSTP096
Gene Symbols C19orf50
Host Species Mouse
Immunogen C19orf50 (NP_076974.1, 1 a.a. ∼ 176 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79036
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.