Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004687 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-687 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-687 Supplier Abnova Corporation Supplier No. H00056913A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant C1GALT1.

The common core 1 O-glycan structure Gal-beta-1-3GalNAc-R is a precursor for many extended mucin-type O-glycan structures in animal cell surface and secreted glycoproteins. Core 1 is synthesized by the transfer of Gal from UDP-Gal to GalNAc-alpha-1-R by core 1 beta-3-galactosyltransferase (C1GALT1) (Ju et al., 2002 [PubMed 11677243]).[supplied by OMIM

Sequence: ETFHPFVPEHHLIKGYLPRTFWYWNYNYYPPVEGPGCCSDLAVSFHYVDSTTMYELEYLVYHLRPYGYLYRYQPTLPERILKEISQANKNEDTKVKLGNP

Specifications

Antigen core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant C1GALT1.
Formulation 50% glycerol
Gene C1GALT1
Gene Accession No. NM_020156
Gene Alias C1GALT/T-synthase
Gene Symbols C1GALT1
Host Species Mouse
Immunogen C1GALT1 (NP_064541, 264 a.a. ∼ 363 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 56913
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.