Learn More
tRNA splicing endonuclease 15 homolog (S. cerevisiae), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human C1orf19 protein.
Supplier: Abnova Corporation H00116461D01P
Description
tRNA splicing is a fundamental process required for cell growth and division. SEN15 is a subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns, the first step in tRNA splicing (Paushkin et al., 2004 [PubMed 15109492]).[supplied by OMIM
Sequence: MEERGDSEPTPGCSGLGPGGVRGFGDGGGAPSWAPEDAWMGTHPKYLEMMELDIGDATQVYVAFLVYLDLMESKSWHEVNCVGLPELQLICLVGTEIEGEGLQTVVPTPITASLSHNRIREILKASRKLQGDPDLPMSFTLAIVESDSTIVYYKLTDGFMLPDPQNISLRRSpecifications
tRNA splicing endonuclease 15 homolog (S. cerevisiae) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human C1orf19 protein. | |
TSEN15 | |
C1orf19/sen15 | |
Rabbit | |
Affinity Purified | |
RUO | |
116461 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_052965.1 | |
TSEN15 | |
C1orf19 (NP_443197.1, 1 a.a. ∼ 171 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.