Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SET domain containing 4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007724 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-724 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-724 Supplier Abnova Corporation Supplier No. H00054093B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human C21orf18 protein.

Sequence: MQKGKGRTSRIRRRKLCGSSESRGVNESHKSEFIELRKWLKARKFQDSNLAPACFPGTGRGLMSQTSLQEGQMIISLPESCLLTTDTVIRSYLGAYITKWKPPPSPLLALCTFLVSEKHAGHRSLWKPYLEILPKAYTCPVCLEPEVVNLLPKSLKAKAEEQRAHVQEFFASSRDFFSSLQPLFAEAVDSIFSYSALLWAWCTVNTRAVYLRPRQRECLSAEPDTCALAPYLDLLNHSPHVQVKAAFNEETHSYEIRTTSRWRKHEEVFICYGPHDNQRLFLEYGFVSVHNPHACVYVSRGWNQLCS

Specifications

Antigen SET domain containing 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human C21orf18 protein.
Formulation No additive
Gene SETD4
Gene Accession No. NM_001007259.1
Gene Alias C21orf18/C21orf27
Gene Symbols SETD4
Host Species Mouse
Immunogen C21orf18 (NP_001007260.1, 1 a.a. ∼ 307 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 54093
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.