Learn More
C21orf66, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human C21orf66 protein.
Supplier: Abnova Corporation H00094104B01P
Description
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq
Sequence: MFRKARRVNVRKRNDSEEEERERDEEQEPPPLLPPPGTGEEAGPGGGDRAPGGESLLGPGPSPPSALTPGLGAEAGGGFPGGAEPGNGLKPRKRPRENKEVPRASLLSFQDEEEENEEVFKVKKSSYSKKIVKLLKKEYKEDLEKSKIKTELNSSAESEQPLDKTGHVKDTNQEDGVIISEHGEDEMDMESEKEEEKPKTGGAFSNALSSLNVLRPGEIPDAAFIHAARKKRQMARELGDFTPHDNEPGKGRLVREDENDASDDEDDDEKRRIVFSVKEKSQRQKIAEEIGIEGSDDDALVTGEQDEELSRWEQEQIRKGINIPQVQASQPAEVNMYYQNTYQTMPYGSSYGIPYSYTAYGSSDAKSQKTDNTVPFKTPSNEMTPVTIDLVKKQLKDRSpecifications
C21orf66 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human C21orf66 protein. | |
C21orf66 | |
BM-020/FLJ90561/GCFC/fSAP105 | |
Mouse | |
Affinity chromatography | |
RUO | |
94104 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC030539.1 | |
C21orf66 | |
C21orf66 (AAH30539.1, 1 a.a. ∼ 398 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.