Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

centromere protein M, Mouse, Clone: 4C12-2C8, Abnova™

Catalog No. 89014334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-334 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-334 Supplier Abnova Corporation Supplier No. H00079019M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant C22orf18.

The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPM is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM

Sequence: MSVLRPLDKLPGLNTATILLVGTEDALLQQLADSMLKEDCASELKVHLAKSLPLPSSVNRPRIDLIVFVVNLHSKYSLQNTEESLRHVDASFFLGKVCFLATGAGRESHCSIHRHTVVKLAHTYQSPLLYCDLEVEGFRATMAQRLVRVLQICAGHVPGVSALNLLSLLRSSEGPSLEDL

Specifications

Antigen centromere protein M
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4C12-2C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant C22orf18.
Formulation PBS with no preservative; pH 7.4
Gene CENPM
Gene Accession No. BC000705
Gene Alias C22orf18/CENP-M/MGC861/PANE1/bK250D10.2
Gene Symbols CENPM
Host Species Mouse
Immunogen C22orf18 (AAH00705, 1 a.a. ∼ 180 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79019
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.