Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3a Rat anti-Mouse, Unlabeled, Clone: I87-1162, BD

Catalog No. BDB558250 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558250 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558250 Supplier BD Biosciences Supplier No. 558250
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The purified I87-1162 antibody reacts with the mouse C3a protein. The I87-1162 antibody is specific for a neoepitope exposed in mouse C3a/C3adesArg and does not cross-react with mouse C3. This capacity makes antibody I87-1162 a preferential capture antibody for direct detection of mouse C3a from plasma or serum samples in an ELISA sandwich assay. Anaphylatoxin C3a is a bioactive cleavage product released from plasma component C3 during complement activation and is involved in mediation of a variety of cellular immune responses, as well as being a potent pro-inflammatory agent. The release of this cleavage product is a reliable indicator of in vivoor in vitrocomplement activation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C3a
Applications ELISA
Classification Monoclonal
Clone I87-1162
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.