Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3a Receptor Mouse anti-Human, Alexa Fluor 647, Clone: hC3aRZ8, BD

Catalog No. BDB561497 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561497 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561497 Supplier BD Biosciences Supplier No. 561497
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The hC3aRZ8 monoclonal antibody specifically binds to the human C3a Receptor (C3aR). C3aR is a seven-transmembrane glycoprotein, G-protein-coupled receptor that is the specific receptor for C3a anaphylatoxin. The C3aR consists of 482 amino acids forming a single polypeptide chain that is encoded by the C3AR1 gene located on chromosome 12 (location 12p13.31). C3aR are expressed by eosinophils, basophils, neutrophils, dendritic cells, monocytes, macrophages, endothelial cells and some T cells. C3a is a bioactive cleavage product released from Complement Component 3 (C3) during complement activation. C3a plays a role in a variety of cellular immune responses as well as being a potent pro-inflammatory agent. In response to bound C3a, this receptor stimulates cellular responses including chemotaxis, granule enzyme release and superoxide anion production and causes increased vascular permeability. In vivo, C3a production can initiate, contribute to, or exacerbate the inflammatory reactions seen in gram-negative bacterial sepsis, trauma, ARDS, ischemic heart disease, post-dialysis syndrome, and several autoimmune diseases including rheumatoid arthritis, lupus erythematosus, and acute glomerulonephritis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C3a Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone hC3aRZ8
Conjugate Alexa Fluor 647
Description C3AR; C3a-R; C3AR1; C3a anaphylatoxin chemotactic receptor; AZ3B
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.