Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4BPB, Mouse, Clone: 1G9, Abnova™

Catalog No. 89016381 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-381 Supplier Abnova Corporation Supplier No. H00000725M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant C4BPB.

This gene encodes a member of a superfamily of proteins composed predominantly of tandemly arrayed short consensus repeats of approximately 60 amino acids. A single, unique beta-chain encoded by this gene assembles with seven identical alpha-chains into the predominant isoform of C4b-binding protein, a multimeric protein that controls activation of the complement cascade through the classical pathway. C4b-binding protein has a regulatory role in the coagulation system also, mediated through the beta-chain binding of protein S, a vitamin K-dependent protein that serves as a cofactor of activated protein C. The genes encoding both alpha and beta chains are located adjacent to each other on human chromosome 1 in the regulator of complement activation gene cluster. Alternative splicing gives rise to multiple transcript variants. [provided by RefSeq

Sequence: MFFWCACCLMVAWRVSASDEHCPELPPVDNSIFVAKEVEGQILGTYVCIKGYHLVGKKTLFCNASKEWDNTTTECRLGHCPDPVLVNGEFSSSGPVNVSDKITFMCNDHYILKGSNRSQCLEDHTWAPPFPICKSRDCDPPGNPVHGYFEGNNFTLGSTISYYCEDRYYLVGVQEQQCVDGEWSSALPVCKLIQEAPKPECEKALLAFQESKNLCEAMENFMQQLKESGMTMEELKYSLELKKAELKAKLL

Specifications

Antigen C4BPB
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant C4BPB.
Formulation PBS with no preservative; pH 7.4
Gene C4BPB
Gene Accession No. BC005378
Gene Alias C4BP
Gene Symbols C4BPB
Host Species Mouse
Immunogen C4BPB (AAH05378, 1 a.a. ∼ 251 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 725
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.