Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4d Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABF11MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF11MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF11MI Supplier MilliporeSigma Supplier No. ABF11

Rabbit Polyclonal Antibody

This Anti-C4d Antibody is validated for use in western blotting for the detection of C4d.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C4d
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols CPAMD3; C4B5; CO4; C4F; C4B2; C4B1; CH; C4B12; C4B3; C4b
Host Species Rabbit
Immunogen Linear peptide corresponding to mouse C4d.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_033910
Target Species Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.