Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement C9 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA519826
Description
For Western Blot, this antibody has non-specific bands at 37 kDa. This antibody is predicted to react with mouse, rat and cow based on sequence homology.
This gene encodes the final component of the complement system. It participates in the formation of the Membrane Attack Complex (MAC). The MAC assembles on bacterial membranes to form a pore, permitting disruption of bacterial membrane organization. Mutations in this gene cause component C9 deficiency.
Specifications
Complement C9 | |
Polyclonal | |
Unconjugated | |
P02748 | |
C9 | |
Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human C9. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
C9 | |
Complement component C9, Membrane Attack Complex component, C5b-9 complex component | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction