Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

family with sequence similarity 13, member B1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-635 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-635 Supplier Abnova Corporation Supplier No. H00051306B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human C5orf5 protein.

Sequence: MRKSSSPSLSNCNSVLANKIFGIPLDELQQGGHPDNEVPFIVRHVVDYIEEHGGLEQQGLFQVNGNAETVEWLRQRYDSGEEVDLVKEADVPSAISLLRFFLQELPEPVIPGSLHIHLMQLSQDYNNEDEFGRKLRFLLQQLPPVNYSLLKFLCRFLANVASHHEEIWSANSLAAVFGPDVFHIYTDVEDMKEQEIVSRIMAGLLENYYEFFENEEEDFSSNDLSSITEQVNELSEEEEEDEKLEHIEELPEEGAEKSNDMPEVVQLRMTENILESNSVTATSTHISPISILPASTDILERTIRAAVEQHLFDLQSSIDHDLKNLQQQSVVCNNEAESIHCDGEGSNNQIDIADDIINASESNRDCSKPVASTNLDNEAMQQDCVFENEENTQSVGILLEPCSDRGDSEDGCLEREEYLLFDSDKLSHLILDSSSKICDLNANTESEVPGGQSVGVQGEAACVSIPHLDLKNVSDGDKWEEPFPAFKSWQEDSESGEAQLSPQAGRMNHHPLEEDCPPVLSHRSLDFGQSQRFLHDPEKLDSSSKALSFTRIRRSSFSSKDEKREDRTPYQLVKKLQKKIRQFEEQFERERNSKPSYSDIAANPKVLKWMTELTKLRKQIKDAKHKNSDGEFVPQTRPRSNTLPKSFGSSLDHEDEENEDEPKVIQKEKKPSKEATLELILKRLKEKRIERCLPEDIKKMTKDHLVEEKASLQKSLLYYESQHGRPVTKEERHIVKPLYDRYRLVKQMLTRASITPVLGSPSTKRRGQMLQPIIEGETAHFFEEIKEEEEDGVNLSSELGDMLKTAVQVQSSLENSESDVEENQEKLALDLRLSSSRAASMPELLEQLWKARAEKKKLRKTLREFEEAFYQQNGRNAQKEDRVPVLEEYREYKKIKAKLRLLEVLISKQDSSKSI

Specifications

Antigen family with sequence similarity 13, member B1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human C5orf5 protein.
Formulation No additive
Gene FAM13B1
Gene Accession No. BC056887.1
Gene Alias C5orf5/DKFZp667F249/FLJ26735/KHCHP/MGC57159/N61
Gene Symbols FAM13B1
Host Species Mouse
Immunogen C5orf5 (AAH56887.1, 1 a.a. ∼ 915 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51306
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.