Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf70 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524511
Description
This antibody is predicted to react with non-human primate based on sequence homology.
Specifications
| C6orf70 | |
| Polyclonal | |
| ERMARD | |
| dJ266L20.3, transmembrane protein C6orf70 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55780 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q5T6L9 | |
| ERMARD | |
| KLH conjugated synthetic peptide between 346-375 amino acids from the central region of human C6orf70 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction