Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chromosome 7 open reading frame 38, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018694 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-694 Supplier Abnova Corporation Supplier No. H00221786D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human C7orf38 protein.

This gene encodes a protein of unknown function. The protein is weakly similar to transposase-like proteins in human and mouse. [provided by RefSeq

Sequence: MTPESRDTTDLSPGGTQEMEGIVIVKVEEEDEEDHFQKERNKVESSPQVLSRSTTMNERALLSSYLVAYRVAKEKMAHTAAEKIILPACMDMVRTIFDDKSADKLRTIPLSDNTISRRICTIAKHLEAMLITRLQSGIDFAIQLDESTDIASCPTLLVYVRYVWQDDFVEDLLCCLNLNSHITGLDLFTELENCLLGQYKLNWKHCKGISSDGTANMTGKHSRLTEKLLEATHNNAVWNHCFIHREALVSKEISPSLMDVLKNAVKTVNFIKGSSLNSRLLEIFCSEIGVNHTHLLFHTEVRWLSQGKVLSRVYELRNEIYIFLVEKQSHLANIFEDDIWVTKLAYLSDIFGILNELSLKMQGKNNDIFQYLEHILGFQKTLLLWQARLKSNRPSYYMFPTLLQHIEENIINEDCLKEIKLEILLHLTSLSQTFNYYFPEEKFESLKENIWMKDPFAFQNPESIIELNLEPEEENELLQLSSSFTLKNYYKILSLSAFWIKIKDDFPLLSRKSILLLLPFTTTYLCELGFSILTRLKTKKRNRLNSAPDMRVALSSCVPDWKELMNRQAHPSH

Specifications

Antigen chromosome 7 open reading frame 38
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human C7orf38 protein.
Formulation PBS with no preservative; pH 7.4
Gene C7orf38
Gene Accession No. NM_145111.2
Gene Alias DKFZp727G131/FLJ36794
Gene Symbols C7orf38
Host Species Rabbit
Immunogen C7orf38 (NP_659802.1, 1 a.a. ∼ 573 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221786
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.