Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C8orf1, Mouse, Clone: 3E11, Abnova™

Catalog No. 89016385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-385 Supplier Abnova Corporation Supplier No. H00000734M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant C8orf1.

Sequence: SGLKKIFKLSAAVVLIGSHPNLSFLKDQGCYLGHKSSQPITCKGNPVEIDTYTYECIKEANLFALGPLVGDNFVRFLKGGALGVTRCLATRQKKKHLFVERGGGDGIA

Specifications

Antigen C8orf1
Applications ELISA
Classification Monoclonal
Clone 3E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant C8orf1.
Formulation PBS with no preservative; pH 7.4
Gene OSGIN2
Gene Accession No. BC031054
Gene Alias C8orf1/hT41
Gene Symbols OSGIN2
Host Species Mouse
Immunogen C8orf1 (AAH31054, 398 a.a. ∼ 505 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 734
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.