Learn More
Ca++ Sensing Receptor Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Ca++ Sensing Receptor |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA |
| Gene Symbols | CASR; PCAR1; CAR; MGC138441; FIH; HHC1; NSHPT; HHC; GPRC2A; FHH; CaSR |
| Host Species | Rabbit |
| Immunogen | A 20 amino acid peptide sequence near the C-terminus of human CaSR. The immunogen sequence is 90% conserved in rat/mouse and 85% in bovine. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.