Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CA095 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525426
Description
C1ORF95 belongs to the SPEC3 family. The exact function of C1ORF95 remains unknown.
Specifications
| CA095 | |
| Polyclonal | |
| Unconjugated | |
| Q69YW2, Q0VBF8 | |
| STUM | |
| KLH conjugated synthetic peptide between 114-141 amino acids from the C-terminal region of human CA095 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| STUM | |
| uncharacterized membrane protein C1orf95 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 375057, 381310 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction