Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calcium channel, voltage-dependent, gamma subunit 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016737 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-737 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-737 Supplier Abnova Corporation Supplier No. H00000786B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CACNG1 protein.

L-type calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of several gamma subunit proteins. This particular gamma subunit is part of skeletal muscle 1,4-dihydropyridine-sensitive calcium channels and is an integral membrane protein that plays a role in excitation-contraction coupling. This gene is a member of the neuronal calcium channel gamma subunit gene subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two similar gamma subunit-encoding genes. [provided by RefSeq

Sequence: MSQTKMLKVRVTLFCILAGIVLAMTAVVTDHWAVLSPHMEHHNTTCEAAHFGLWRICTKRIPMDDSKTCGPITLPGEKNCSYFRHFNPGESSEIFEFTTQKEYSISAAAIAIFSLGFIILGSLCVLLSLGKKRDYLLRPASMFYAFAGLCILVSVEVMRQSVKRMIDSEDTVWIEYYYSWSFACACAAFILLFLGGLALLLFSLPRMPRNPWESCMDAEPEH

Specifications

Antigen calcium channel, voltage-dependent, gamma subunit 1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CACNG1 protein.
Formulation PBS with no preservative; pH 7.4
Gene CACNG1
Gene Accession No. NM_000727
Gene Alias CACNLG
Gene Symbols CACNG1
Host Species Mouse
Immunogen CACNG1 (NP_000718.1, 1 a.a. ∼ 222 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 786
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.