Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNG7, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89203983 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-203-983 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-203-983 Supplier Abnova Corporation Supplier No. H00059284K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human CACNG7 peptide using ARM Technology.

The mouse protein stargazin is one of five subunits comprising neuronal voltage-gated calcium channels. This subunit, gamma, is thought to stabilize the calcium channel in an inactive (closed) state. Mutations in the gene encoding stargazin have been associated with absence seizures, also known as petit-mal or spike-wave seizures. The protein encoded by this gene is structurally similar to the mouse stargazin protein and is a member of the neuronal calcium channel gamma subunit protein family. However, it appears unlikely that the encoded protein is part of a functional calcium channel. Rather, it appears to inhibit the expression of a specific calcium channel subunit. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen CACNG7
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description calcium channel, voltage-dependent, gamma subunit 7
Formulation In 1X PBS, pH 7.4
Gene CACNG7
Gene Accession No. CACNG7
Gene Symbols CACNG7
Host Species Rabbit
Immunogen A synthetic peptide of human CACNG7
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59284
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.