Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Cadherin-13, APC, Clone: 392411, R&D Systems™

Catalog No. FAB3264A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3264A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3264A Supplier R&D Systems Supplier No. FAB3264A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen Cadherin-13
Applications Flow Cytometry
Classification Monoclonal
Clone 392411
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P55290
Gene Alias cadherin 13, H-cadherin (heart), cadherin-13, Cadherin13, CDH13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, T-Cadherin, Truncated cadherin, truncated-cadherin
Gene Symbols CDH13
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cadherin-13, Glu23-Ala692, Accession # P55290
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1012
Test Specificity Detects human Cadherin-13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) E-Cadherin, rhN-Cadherin, rhP-Cadherin, rhVE-Cadherin, rhCadherin-8, -11, -12 or -17 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.