Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-5 Mouse anti-Human, Unlabeled, Clone: 75, BD

Catalog No. BDB610251 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610251 50 μg
BDB610252 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610251 Supplier BD Biosciences Supplier No. 610251
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherins are a family of transmembrane glycoproteins involved in the Ca2+- dependent cell-cell adhesion that occurs in many tissues. These proteins are similar in their domain structure (45-74% amino acid conservation), Ca2+ and protease-sensitivity, and molecular weight. Cadherin-5 (VE-Cadherin or CD144) is one of a number of cadherins (cadherin-4 through -11) whose cDNAs were isolated from rat brain and retina. These cadherins have a cytoplasmic domain that is highly conserved relative to previously identified cadherins, indicating that this domain is essential for cell adhesion activity. This function is mediated by cadherin interaction with cytoskeletal proteins. However, Cadherin-5's cytoplasmic domain has the lowest degree of homology with the other cadherins. Cadherin-5 is expressed in brain and various other tissues, including umbilical cord vein endothelial cells. A new type of adhering junction has been identified in certain vascular endothelial cells. These junctions are known as “complexus adherens” and are morphologically and compositionally distinct from desmosomes and zonula adherens junctions. The complexus adherens of endothelial cells lack desmosomal cadherins as well as E-Cadherin. However, these cells are rich in Cadherin-5 which colocalizes with desmoplakin and γ-Catenin (plakoglobin).

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cadherin-5
Applications Western Blot
Classification Monoclonal
Clone 75
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.