Learn More
Ca++-dependent secretion activator, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CADPS.
Supplier: Abnova Corporation H00008618A01
Description
This gene encodes a novel neural/endocrine-specific cytosolic and peripheral membrane protein required for the Ca2+-regulated exocytosis of secretory vesicles. The protein acts at a stage in exocytosis that follows ATP-dependent priming, which involves the essential synthesis of phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2). Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described. [provided by RefSeq]
Sequence: RLFDQWYNSSMNVICTWLTDRMDLQLHIYQLKTLIRMVKKTYRDFRLQGVLDSTLNSKTYETIRNRLTVEEATASVSEGGGLQGISMKDSDEEDEEDDSpecifications
| Ca++-dependent secretion activator | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CADPS. | |
| CADPS | |
| CADPS1/CAPS/CAPS1/KIAA1121 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 8618 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003716 | |
| CADPS | |
| CADPS (NP_003707, 1256 a.a. ∼ 1353 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.