Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calbindin 1, 28kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017466 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-466 Supplier Abnova Corporation Supplier No. H00000793D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CALB1 protein.

Calbindin is a calcium-binding protein belonging to the troponin C superfamily. It was originally described as a 27kDa protein induced by vitamin D in the duodenum of the chick. In the brain, its synthesis is independent of vitamin-D-derived hormones. Calbindin contains 4 active calcium-binding domains, and 2 modified domains that presumably have lost their calcium-binding capacity. The neurons in brains of patients with Huntington disease are calbindin-depleted. [provided by RefSeq

Sequence: MAESHLQSSLITASQFFEIWLHFDADGSGYLEGKELQNLIQELQQARKKAGLELSPEMKTFVDQYGQRDDGKIGIVELAHVLPTEENFLLLFRCQQLKSCEEFMKTWRKYDTDHSGFIETEELKNFLKDLLEKANKTVDDTKLAEYTDLMLKLFDSNNDGKLELTEMARLLPVQENFLLKFQGIKMCGKEFNKAFELYDQDGNGYIDENELDALLKDLCEKNKQDLDINNITTYKKNIMALSDGGKLYRTDLALILCAGDN

Specifications

Antigen calbindin 1, 28kDa
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CALB1 protein.
Formulation PBS with no preservative; pH 7.4
Gene CALB1
Gene Accession No. NM_004929
Gene Alias CALB
Gene Symbols CALB1
Host Species Rabbit
Immunogen CALB1 (NP_004920.1, 1 a.a. ∼ 261 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 793
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.