Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Calbindin D-28K (Ab-1), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC253L100UL
Description
Specifically detects Calbindin D-28K Clone: in Baboon, Human, Monkey, Rat, Virus samples, and it is validated for Immunohistochemistry (Frozen)
Several EF-hand calcium-binding proteins have been identified within the nervous system, which includes calbindin, calretinin, and parvalbumin. These cytosolic calcium-binding proteins modulate and mediate the function of the intracellular second messenger calcium. These proteins are believed to function in the buffering, transport and regulation of calcium. Calbindin, calretinin, and parvalbumin are localized in distinct neuronal subpopulations, and antibodies against these proteins are useful markers for neuranatomical and developmental studies.
Specifications
Calbindin D-28K | |
Polyclonal | |
Undiluted serum. | |
purified bovine cerebellum calbindin D-28K protein | |
RUO | |
Recognizes the Calbindin D-28K protein in rat hippocampus tissue. | |
−20°C | |
IgG |
Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
Baboon, Human, Monkey, Rat, Virus | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction