Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin Mouse anti-Human, Unlabeled, Clone: G182-1847, BD

Catalog No. BDB556350 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556350 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556350 Supplier BD Biosciences Supplier No. 556350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calcineurin is a Ca2+- and calmodulin-dependent serine/threonine phosphatase which consists of two subunits. The A subunit (∽61 kDa) is catalytic and has calmodulinbinding activity; the B subunit (∽19 kDa), which is myristylated at its N-terminus, has Ca[2+]-binding activity. Calcineurin is expressed ubiquitously in eukaryotic cells, including yeast. The protein is highly expressed in mammalian brain, and has been implicated in a number of calcium-dependent pathways in the nervous system, including regulation of neurotransmitter release. Calcineurin also plays a key role in T cell receptor-mediated signal transduction pathways. Calcineurin dephosphorylates components of NF-AT (Nuclear Factor of Activated T cells), allowing NF-AT to translocate to the nucleus where it participates with AP-1 in activation of multiple cytokine and surface receptor genes. Inhibition of calcineurin by immunosuppressive agents such as cyclosporin A and FK506 correlates with the suppression of early T cell activation events. These immunosuppressive agents bind to specific intracellular proteins, called immunophilins, to form complexes that inhibit calcineurin’s phosphatase activity. In the presence of these complexes, NF-AT remains phosphorylated and thus cannot translocate to the nucleus. A novel protein, Cain (calcineurin inhibitor), has been described which displays a similar pattern of neuronal and brain expression and has been shown to bind and inhibit calcineurin activity in vivo.

G182-1847 recognizes the α and β isoforms of the A subunit of calcineurin (∼61 kDa). It reacts with an epitope in the autoinhibitory domain. It does not recognize the B subunit of calcineurin. A recombinant tag-fusion protein containing amino acids 349-505 of the human β1 isoform of calcineurin A subunit was used as immunogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcineurin
Applications Western Blot
Classification Monoclonal
Clone G182-1847
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human β1 isoform of calcineurin A Subunit aa. 349-505
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.