Learn More
Description
Specifications
Specifications
| Antigen | Calcineurin pan A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 40% gylerol. Contains no preservative. |
| Gene Symbols | CALN; CALNA; CALNA1; CCN1 ; CNA; CNA1; EC 3.1.3.16; PPP2B |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated synthetic peptide corresponding to Calcineurin. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.