Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Calcineurin/PP2B Aα, Polyclonal,

Catalog No. 07067MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-067-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-067-MI Supplier MilliporeSigma™ Supplier No. 07067

Specifications

Antigen Calcineurin/PP2B Aα
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, 0.15M NaCl, 0.05% sodium azide prior to the addition of glycerol to 30%
Gene Symbols PPP3CA; CALN; CALNA; CCN1; CNA1; CALNA1; PPP2B; CNA
Host Species Rabbit
Immunogen 25 residue, synthetic peptide corresponding to amino acids 490-514 (MPSDANLNSINKALASETNGTDSNG) of bovine Calcineurin/PP2B Aα
Purification Method Ammonium sulfate precipitation
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_008913
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.