Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Calcineurin/PP2B, Clone: VA1,

Catalog No. 05187MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-187-MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-187-MI Supplier MilliporeSigma™ Supplier No. 05187

Specifications

Antigen Calcineurin/PP2B
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone VA1
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, containing 0.05% sodium azide
Gene Accession No. NM_000945.3
Gene Symbols PPP3R1, CNB1, CALNB1, CNB, CNA2
Host Species Mouse
Immunogen Purified bovine PP2B
Purification Method DEAE chromatography, Affinity Gel blue
Quantity 50 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.