Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcitonin Gene Related Peptide Mouse anti-Guinea Pig, Human, Clone: CD8, MilliporeSigma™

Catalog No. MAB317MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB317MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB317MI Supplier MilliporeSigma Supplier No. MAB317
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Calcitonin Gene Related Peptide Antibody, α hCGRP a.a. 28-37, clone CD8 is an antibody against Calcitonin Gene Related Peptide for use in ELISA, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcitonin Gene Related Peptide
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone CD8
Conjugate Unconjugated
Gene Accession No. NM_001033952.1; NM_001033953.1; NM_001741.2
Gene Symbols CALCA, CGRP-I, katacalcin, KC, CALC1, CGRP, MGC126648, calcitonin, CT, CGRP1
Host Species Mouse
Immunogen A terminal fragment of hCGRP coupled to BSA with glutaraldehyde.
Quantity 50 μg
Primary or Secondary Primary
Target Species Guinea Pig, Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.