Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Calcium/Calmodulin Kinase I, Clone: EPR2217Y, Rabbit Monoclonal,

Catalog No. 041158MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-115-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-115-8MI Supplier MilliporeSigma™ Supplier No. 041158

Specifications

Antigen Calcium/Calmodulin Kinase I
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone EPR2217Y
Conjugate Unconjugated
Formulation Rabbit Monoclonal in buffer containing 50mM Tris-Glycine (pH 7.4), 0.15M NaCl containing 40% Glycerol, 0.01% sodium azide and 0.05% BSA.
Gene Accession No. NP_003647.1
Host Species Rabbit
Immunogen Specific peptide corresponding to residues on the N-terminus of human Calcium/Calmodulin Kinase I.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) CAMKK, CAMKKB, EC 2.7.11.17, KIAA0787, MGC15254
Target Species Human, Mouse, Rat
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.