Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calcium binding and coiled-coil domain 2, Mouse, Clone: 2A11, Abnova™

Catalog No. 89001170 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-170 Supplier Abnova Corporation Supplier No. H00010241M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CALCOCO2.

The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles. [provided by RefSeq

Sequence: SYMGLDFNSLPYQVPTSDEGGARQNPGLAYGNPYSGIQESSSPSPLSIKKCPICKADDICDHTLEQQQMQPLCFNCPICDKIFPATEKQIFEDHVFCHSL

Specifications

Antigen calcium binding and coiled-coil domain 2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2A11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CALCOCO2.
Formulation PBS with no preservative; pH 7.4
Gene CALCOCO2
Gene Accession No. NM_005831
Gene Alias MGC17318/NDP52
Gene Symbols CALCOCO2
Host Species Mouse
Immunogen CALCOCO2 (NP_005822, 347 a.a. ∼ 446 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10241
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.