Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALCR, Mouse, Clone: 2F7, Abnova™

Catalog No. 89016408 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-408 Supplier Abnova Corporation Supplier No. H00000799M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CALCR.

This gene encodes a high affinity receptor for the peptide hormone calcitonin and belongs to a subfamily of seven transmembrane-spanning G protein-coupled receptors. The encoded protein is involved in maintaining calcium homeostasis and in regulating osteoclast-mediated bone resorption. Polymorphisms in this gene have been associated with variations in bone mineral density and onset of osteoporosis. Alternate splicing results in multiple transcript variants. [provided by RefSeq

Sequence: CNNEVQTTVKRQWAQFKIQWNQRWGRRPSNRSARAAAAAAEAGDIPIYICHQELRNEPANNQGEESAEIIPLNIIEQESSA

Specifications

Antigen CALCR
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CALCR.
Formulation PBS with no preservative; pH 7.4
Gene CALCR
Gene Accession No. NM_001742
Gene Alias CRT/CTR/CTR1
Gene Symbols CALCR
Host Species Mouse
Immunogen CALCR (NP_001733.1, 394 a.a. ∼ 474 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 799
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.