Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcyclin Mouse anti-Human, Clone: CPTC-Calcyclin-2, MilliporeSigma™

Catalog No. MABS31MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS31MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS31MI Supplier MilliporeSigma Supplier No. MABS31
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Calcyclin Antibody, clone CPTC-Calcyclin-2 detects level of Calcyclin and has been published and validated for use in western blot, ELISA, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcyclin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone CPTC-Calcyclin-2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_055439
Gene Symbols MLN 4, PRA, CACY, CABP, 2A9, 5B10
Host Species Mouse
Immunogen Full length recombinant protein corresponding to human Calcyclin.

Material obtained from the National Cancer Institute's Clinical Proteomic Technologies for Cancer (CPTC) program.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.