Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALML3, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002880 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-880 Supplier Abnova Corporation Supplier No. H00000810D01
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CALML3 protein.

Sequence: MADQLTEEQVTEFKEAFSLFDKDGDGCITTRELGTVMRSLGQNPTEAELRDMMSEIDRDGNGTVDFPEFLGMMARKMKDTDNEEEIREAFRVFDKDGNGFVSAAELRHVMTRLGEKLSDEEVDEMIRAADTDGDGQVNYEEFVRVLVSK

Specifications

Antigen CALML3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CALML3 protein.
Formulation No additive
Gene CALML3
Gene Accession No. NM_005185.2
Gene Alias CLP
Gene Symbols CALML3
Host Species Rabbit
Immunogen CALML3 (NP_005176.1, 1 a.a. ∼ 149 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 810
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.