Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Calmodulin Ab-4, Clone: 1F11 + 2D1 + 6D4, Epredia™ LabVision

Catalog No. MS1268P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
0.5 mL
1 mL
Concentration:
1mg/mL
200μg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1268P1 0.5 mL 200μg/mL
MS1268P 1 mL 200μg/mL
MS1268P1ABX 100 μL 1mg/mL
MS1268PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1268P1 Supplier Epredia Supplier No. MS1268P1

Mouse Monoclonal Antibody

Calmodulin (CaM) is a calcium-binding protein found in all eukaryotic cells. It belongs to a structurally homologous family of proteins which includes troponin C, parvalbumin, and S-100. CaM is highly conserved among species, and all vertebrate CaMs have identical structures. Although CaM exists in abundance in many cells, it can be regulated by hormones and neurotransmitters in a variety of tissues and systems.

Host Species: Mouse
Clone: 1F11 + 2D1 + 6D4
Isotype: IgG1 + IgG1 + IgG1
Cross Reactivity: Human, Cow, Rat, Chicken, and Dictyostelium discoideum. Others not tested.
Epitope: Not determined
Immunogen: Purified calmodulin from Dictyostelium discoideum strain.
Molecular Weight: 17kDa
Positive Control: Dictyostelium lysate on IMR5 cells.
Cellular Localization: Cytoplasmic

Ab-4 is a cocktail of Ab-1, Ab-2, and Ab-3.

Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen Calmodulin
Classification Monoclonal
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 0.5 mL
Primary or Secondary Primary
Target Species Chicken, Human, Rat
Form Purified
Isotype IgG1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.